Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadherin-17 Antibody (CDH17/2618), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279857APC
Description
Cadherin-17 Monoclonal specifically detects Cadherin-17 in Human samples. It is validated for Flow Cytometry.Specifications
Cadherin-17 | |
Monoclonal | |
APC | |
PBS with 0.05% Sodium Azide | |
CDH17 | |
Recombinant fragment (around aa 242-418) of human Cadherin 17 protein (CDH17) (exact sequence is proprietary) (Uniprot: Q12864) | |
0.1 mL | |
Cancer | |
This antibody recognizes a protein of 120kDa, which is identified as Cadherin-17 (also known as LI Cadherin). | |
Store at 4°C in the dark. |
Flow Cytometry | |
CDH17/2618 | |
Flow Cytometry | |
cadherin, cadherin 17, LI cadherin (liver-intestine), cadherin-16, cadherin-17, CDH16, FLJ26931, HPT1, HPT-1, HPT-1 cadherin, human intestinal peptide-associated transporter HPT-1, human peptide transporter 1, Intestinal peptide-associated transporter HPT-1, LI cadherin, LI-cadherin, Liver-intestine cadherin, MGC138218, MGC142024 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction