Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadherin-17 Antibody (rCDH17/8514), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP320414100UG
Description
Cadherin-17 Monoclonal antibody specifically detects Cadherin-17 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Cadherin-17 | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 μg/mL | |
cadherin, cadherin 17, LI cadherin (liver-intestine), cadherin-16, cadherin-17, CDH16, FLJ26931, HPT1, HPT-1, HPT-1 cadherin, human intestinal peptide-associated transporter HPT-1, human peptide transporter 1, Intestinal peptide-associated transporter HPT-1, LI cadherin, LI-cadherin, Liver-intestine cadherin, MGC138218, MGC142024 | |
Recombinant fragment (around aa 242-418) of human Cadherin-17 protein (CDH17) (exact sequence is proprietary) | |
100 μg | |
Cancer | |
1015 | |
Store at 4&drg;C. Do not freeze. | |
IgG1 κ |
Immunohistochemistry (Paraffin) | |
rCDH17/8514 | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction