Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadherin-4/R-Cadherin Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$181.90 - $402.00

Specifications

Antigen Cadherin-4/R-Cadherin
Concentration 0.5 mg/mL
Dilution Western Blot 1:100-1:2000
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP227372SS
SDP
View Documents
Novus Biologicals
NBP227372SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NBP227372
SDP
View Documents
Novus Biologicals
NBP227372
0.1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

Cadherin-4/R-Cadherin Polyclonal specifically detects Cadherin-4/R-Cadherin in Human, Mouse, Mammal samples. It is validated for Western Blot.
Specifications

Specifications

Cadherin-4/R-Cadherin
Western Blot 1:100-1:2000
Polyclonal
Purified
RUO
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
CAD4, cadherin 4, type 1, preproprotein, cadherin 4, type 1, R-cadherin (retinal), cadherin-4, FLJ22202, FLJ40547, MGC126700, MGC138355, RCAD, R-CAD, R-cadherin, Retinal cadherin
CDH4
IgG
Protein G purified
The amino acid sequence used as immunogen for the antibody is conserved in most mammals.
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
Human, Mouse, Mammalia
P55283
1002
Amino acids DYDYLNDWGPRFkkLADMYGGGDD of human Cadherin was used as the immunogen for this antibody.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.