Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadherin-6/KCAD Antibody (SR1127), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Cadherin-6/KCAD |
---|---|
Clone | SR1127 |
Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
Cadherin-6/KCAD Monoclonal antibody specifically detects Cadherin-6/KCAD in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
Cadherin-6/KCAD | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
cadherin 6, type 2, K-cadherin (fetal kidney), cadherin, fetal kidney, cadherin-6, KCAD, K-cadherin, K-cadherin (fetal kidney) | |
A synthesized peptide derived from human Cadherin-6/KCAD (Uniprot #: P55285) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1127 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
1004 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title