Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadherin-8 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298434JF646
Description
Cadherin-8 Polyclonal specifically detects Cadherin-8 in Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cadherin-8 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
cadherin 8, type 2, cadherin-8, Nbla04261, putative protein product of Nbla04261 | |
Produced in rabbits immunized with purified, recombinant Cadherin-8 (NP_445845.2; Met1-Met621). | |
0.1 mL | |
Primary | |
Rat | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
1006 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction