Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadherin-8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$330.00 - $547.00
Specifications
Antigen | Cadherin-8 |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Cadherin-8 Polyclonal antibody specifically detects Cadherin-8 in Human samples. It is validated for ImmunofluorescenceSpecifications
Cadherin-8 | |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
PBS, pH 7.2, 40% glycerol | |
1006 | |
IgG | |
Affinity purified |
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
cadherin 8, type 2, cadherin-8, Nbla04261, putative protein product of Nbla04261 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: KDDPKNGHYFLYSLLPEMVNNPNFTIKKNEDNSLSILAKHNGFNRQKQEVYLLPIII | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title