Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadherin-8 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen Cadherin-8
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP159221
SDP
View Documents
Novus Biologicals
NBP159221
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

Cadherin-8 Polyclonal specifically detects Cadherin-8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Cadherin-8
Polyclonal
Rabbit
P55286
1006
Synthetic peptides corresponding to CDH8(cadherin 8, type 2) The peptide sequence was selected from the middle region of CDH8. Peptide sequence HENAALNSVIGQVTARDPDITSSPIRFSIDRHTDLERQFNINADDGKITL.
Primary
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
cadherin 8, type 2, cadherin-8, Nbla04261, putative protein product of Nbla04261
CDH8
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.