Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Cadherin E Ab-4, Mouse Monoclonal Antibody

Catalog No. MS1862R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1862R7 7 mL Prediluted
MS1862S1 500 μL Concentrated
MS1862S0 100 μL Concentrated
MS1862R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS1862R7 Supplier Epredia™ Supplier No. MS1862R7

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cadherin E Ab-4, Mouse Monoclonal Antibody.

E-cadherin (uvomorulin, cell-CAM120/80) is a calcium-dependent cell adhesion molecule expressed predominately in epithelial tissues. It plays an important role in the growth and development of cells via the mechanisms of control of tissue architecture and the maintenance of tissue integrity. Numerous studies have demonstrated that reduction and/or loss of E-cadherin expression in carcinomas correlates positively with the potential of these tumors for invasion and metastasis.

Host Species: Mouse

Clone: NCH-38

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: E-cadherin and GST recombinant protein

Molecular Weight: 120 kDa

Positive Control: Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)

Note:

USA: IVD; Int'l: IVD

Specifications

Antigen Cadherin E/E-Cadherin Ab-4
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone NCH-38
Conjugate Unconjugated
Host Species Mouse
Immunogen E-cadherin and GST recombinant protein
Quantity 7 mL
Regulatory Status RUO
Research Discipline Junction Protein
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.