Learn More
Description
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse, Rat, Chicken, Dog, Monkey, Cow and Xenopus.
Epitope: C-terminal
Immunogen: A synthetic peptide from the C-terminal end of chicken N-cadherin
Positive Control: Tonsil, squamous epithelium
Cellular Localization: Cell membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Cadherin, Pan Ab-4 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Ready-to-use for IHC (UVLP) |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide from the C-terminal end of chicken N-cadherin |
| Purification Method | Tonsil, squamous epithelium |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.