Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CADM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106549
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106549 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106549 Supplier Invitrogen™ Supplier No. PA5106549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CADM1.

Mediates homophilic cell-cell adhesion in a Ca (2+)-independent manner. Also mediates heterophilic cell-cell adhesion with CADM3 and PVRL3 in a Ca (2+)-independent manner. Acts as a tumor suppressor in non-small-cell lung cancer (NSCLC) cells. Interaction with CRTAM promotes natural killer (NK) cell cytotoxicity and interferon-gamma (IFN-gamma) secretion by CD8+ cells in vitro as well as NK cell-mediated rejection of tumors expressing CADM3 in vivo. May contribute to the less invasive phenotypes of lepidic growth tumor cells. In mast cells, may mediate attachment to and promote communication with nerves. CADM1, together with MITF, is essential for development and survival of mast cells in vivo. May act as a synaptic cell adhesion molecule that drives synapse assembly. May be involved in neuronal migration, axon growth, pathfinding, and fasciculation on the axons of differentiating neurons. May play diverse roles in the spermatogenesis including in the adhesion of spermatocytes and spermatids to Sertoli cells and for their normal differentiation into mature spermatozoa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CADM1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CADM1
Gene Accession No. Q8R5M8, Q9BY67
Gene Alias 2900073G06Rik; 3100001I08Rik; AI987920; Bl2; cadm1; Cell adhesion molecule 1; cell adhesion molecule 1 transcript variant 8abc; Igsf4; Igsf4a; Immunoglobulin superfamily member 4; immunoglobulin superfamily member 4A; immunoglobulin superfamily, member 4; immunoglobulin superfamily, member 4A; immunoglobulin superfamily, member 4D variant 1; immunoglobulin superfamily, member 4D variant 2; immunosuperfamily protein Bl2; Necl2; NECL-2; nectin-like 2; nectin-like protein 2; RA175; RA175A; RA175B; RA175C; RA175N; secretory isoform of TSLC1; sgigsf; Spermatogenic immunoglobulin superfamily; ST17; sTSLC-1; Synaptic cell adhesion molecule; SynCAM; SynCam1; truncated CADM1 protein; TSLC1; TSLC-1; TSLC1/Nectin-like 2/IGSF4; tumor suppressor in lung cancer 1; Tumor suppressor in lung cancer 1 homolog
Gene Symbols CADM1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CADM1(Accession Q9BY67), corresponding to amino acid residues S105-T155.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23705, 54725
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.