Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CADM2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549906
Description
The antibody detects endogenous levels of total CADM2 protein.
This gene encodes a member of the synaptic cell adhesion molecule 1 (SynCAM) family which belongs to the immunoglobulin (Ig) superfamily. The encoded protein has three Ig-like domains and a cytosolic protein 4.1 binding site near the C-terminus. Proteins belonging to the protein 4.1 family crosslink spectrin and interact with other cytoskeletal proteins. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
CADM2 | |
Polyclonal | |
Unconjugated | |
CADM2 | |
2900078E11Rik; 9330131D06; A830029E02Rik; Cadm2; Cell adhesion molecule 2; Igdf4d; Igsf4d; immunoglobulin superfamily member 4D; immunoglobulin superfamily, member 4; immunoglobulin superfamily, member 4D; NECL3; Necl-3; nectin-like 3; nectin-like protein 3; Synaptic cell adhesion molecule 2; SynCAM 2; SynCAM2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
239857, 253559, 360687 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q1WIM2, Q8BLQ9, Q8N3J6 | |
CADM2 | |
Synthesized peptide derived from internal of human CADM2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction