Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CADM3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547524
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547524 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547524 Supplier Invitrogen™ Supplier No. PA547524
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) IGSF4C is observed and less than 2% cross-reactivity with rhIGSF4, rhIGSF4D, and rhIGSF8 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Cell adhesion molecule 3 (CADM3), also known as Necl-1, is a gene located on chromosome 1p36.22, encoding a protein that is part of the immunoglobulin superfamily involved in cell-cell adhesion processes. CADM3 is predominantly expressed in the nervous system, particularly in regions such as the brain and spinal cord, where it plays crucial roles in neuronal communication, synapse formation, and maintenance of neural network architecture. The protein functions by mediating homophilic and heterophilic interactions with other cell adhesion molecules, facilitating synapse stabilization and neuronal differentiation. CADM3's activity is essential for proper neural development and function, and dysregulation of its expression has been linked to various neurological disorders. Beyond its role in the nervous system, CADM3 also contributes to tumor suppression in certain cancers, as its expression is often downregulated in tumor tissues. Research into CADM3 continues to explore its potential as a therapeutic target in both neurobiology and oncology.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CADM3
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Cadm3
Gene Accession No. Q8N126
Gene Alias BIgR; brain immunoglobulin receptor; CADM3; Cell adhesion molecule 3; dendritic cell nectin-like protein 1 short isoform; IGSF4B; Immunoglobulin superfamily member 4B; immunoglobulin superfamily, member 4B; NECL1; NECL-1; nectin-like 1; nectin-like protein 1; nectin-lke 1; synaptic cell adhesion molecule 3; SYNCAM3; TSLC1-like 1; TSLC1-like protein 1; TSLL1; UNQ225/PRO258
Gene Symbols Cadm3
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human IGSF4B/SynCAM3 isoform 1 Pro21-Tyr329.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57863
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.