Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CADM3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986986
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6986 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6986 Supplier Bioss Supplier No. BS6027R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IGSF4B is a brain-specific protein related to the calcium-independent cell-cell adhesion molecules known as nectins.

Specifications

Antigen CADM3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Cadm3
Gene Accession No. Q8N126
Gene Alias BIgR; BOS_2658; Brain immunoglobulin receptor; Cadm3; Cell adhesion molecule 3; dendritic cell nectin-like protein 1 short isoform; IGSF4B; Immunoglobulin superfamily member 4B; immunoglobulin superfamily, member 4B; Necl1; NECL-1; nectin-like 1; Nectin-like protein 1; nectin-lke 1; Synaptic cell adhesion molecule 3; Syncam3; TSLC1-like 1; TSLC1-like protein 1; Tsll1; UNQ225/PRO258
Gene Symbols Cadm3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CADM3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57863
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.