Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CADM4 Monoclonal Antibody (404619)
GREENER_CHOICE

Catalog No. PIMA524141
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524141 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524141 Supplier Invitrogen™ Supplier No. MA524141
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This antibody detects recombinant human (rh) IGSF4C/SynCAM4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) IGSF4A, rhIGSF4B, rhIGSF4D, or rhIGSF8 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

CADM4 is involved in the cell-cell adhesion. Has calcium- and magnesium-independent cell-cell adhesion activity. May have tumor-suppressor activity. Fukuhara H., Oncogene 20:5401-5407(2001). Grimwood J., Nature 428:529-535(2004)0. Williams Y.N., Oncogene 25:1446-1453(2006).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CADM4
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 404619
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene CADM4
Gene Accession No. Q8NFZ8
Gene Alias Cadm4; Cell adhesion molecule 4; Igdf4c; IGSF4C; Immunoglobulin superfamily member 4C; immunoglobulin superfamily, member 4C; membrane glycoprotein Tsll2; Necl4; NECL-4; nectin-like 4; nectin-like protein 4; SynCAM 4; synCAM4; TSLC1-like 2; TSLC1-like protein 2; Tsll2; Upar; urokinase plasminogen activator receptor
Gene Symbols CADM4
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human IGSF4C/SynCAM4 Gln25-Tyr323.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 199731
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.