Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CADM4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552958
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552958 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552958 Supplier Invitrogen™ Supplier No. PA552958
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GGIIICEAQN QALPSGHSKQ TQYVLDVQYS PTARIHASQA VVREGDTLVL TCAVTGNPRP NQIRWNRGNE SLPERAEAVG ETLTLPGLVS ADNGTYTCEA SNKHGHARAL YVLVVYDPGA VVEAQTSVPY Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

CADM4 is involved in the cell-cell adhesion. Has calcium- and magnesium-independent cell-cell adhesion activity. May have tumor-suppressor activity. Fukuhara H., Oncogene 20:5401-5407(2001). Grimwood J., Nature 428:529-535(2004)0. Williams Y.N., Oncogene 25:1446-1453(2006).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CADM4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CADM4
Gene Accession No. Q8NFZ8
Gene Alias Cadm4; Cell adhesion molecule 4; Igdf4c; IGSF4C; Immunoglobulin superfamily member 4C; immunoglobulin superfamily, member 4C; membrane glycoprotein Tsll2; Necl4; NECL-4; nectin-like 4; nectin-like protein 4; SynCAM 4; synCAM4; TSLC1-like 2; TSLC1-like protein 2; Tsll2; Upar; urokinase plasminogen activator receptor
Gene Symbols CADM4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CADM4. Recombinant protein control fragment (Product #RP-89894).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 199731
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.