Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CADM4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501734003 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4003 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4003 Supplier Proteintech Group Inc Supplier No. 231361AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CADM4 is involved in the cell-cell adhesion. Has calcium- and magnesium-independent cell-cell adhesion activity. May have tumor-suppressor activity. Fukuhara H., Oncogene 20:5401-5407(2001). Grimwood J., Nature 428:529-535(2004)0. Williams Y.N., Oncogene 25:1446-1453(2006).

Specifications

Antigen CADM4
Applications Western Blot
Classification Polyclonal
Concentration 0.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CADM4
Gene Accession No. Q1WIM1, Q8NFZ8, Q8R464
Gene Alias CADM4, cell adhesion molecule 4, IGSF4C, Necl 4, NECL4, Nectin like protein 4, synCAM4, TSLC1 like protein 2, TSLL2
Gene Symbols Cadm4
Host Species Rabbit
Immunogen CADM4 Fusion Protein Ag18922
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 199731, 260299, 365216
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.