Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium, AAS standard solution, Specpure™ Cd 1000μg/mL

Catalog No. p-7080399
Click to view available options
Quantity:
100 mL
500 mL

Cd in 5% HNO3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula Cd in 5% HNO3
MDL Number MFCD00010914

Specifications

Physical Form Liquid
Concentration Cd in 5% HNO3
UN Number UN3264
Quantity 100 mL
Grade Specpure™
Chemical Name or Material Cadmium, AAS standard solution, Cd 1000μg/mL
Hazard Category H290-H315-H318-H350
Hazard Statement GHS H Statement
H350-H361-H314
May cause cancer.
Suspected of damaging fertility or the unborn child.
Causes severe skin burns and eye damage.
Precautionary Statement P201-P202-P234-P264b-P281-P302+P352-P305+P351+P338-P308+P313-P310-P332+P313-P362-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.