Learn More
Cadmium acetate dihydrate, 98%
Description
Cadmium acetate dihydrate is in the porcelain production for the production of iridescent used effects. Moreover, it is in the analytical chemistry, a reagent for the detection of sulfur, selenium and tellurium. It is also used as chemical and pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | White |
Physical Form | Powder |
Quantity | 500 g |
UN Number | UN2570 |
Merck Index | 14,1614 |
Solubility Information | Freely soluble in water, soluble in alcohol |
Formula Weight | 266.54 (230.50 Anhydrous) |
Percent Purity | 98% |
Sensitivity | Hygroscopic |
Density | 2.01 g/mL |
Show More |
Safety and Handling
Hazard Category | H302+H312+H332-H335-H350-H373 |
Hazard Statement | GHS H Statement H302-H312-H332 Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. |
Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS |
EINECSNumber | 208-853-2 |
RTECSNumber | AF7505000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.