Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium acetate dihydrate, 98%, for analysis

Catalog No. p-3756619
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle

CAS: 5743-04-4 | C4H6CdO4·2H2O | 266.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5743-04-4
Molecular Formula C4H6CdO4·2H2O
Molecular Weight (g/mol) 266.52
MDL Number MFCD00150020
InChI Key AUIZLSZEDUYGDE-UHFFFAOYSA-L

Specifications

Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 100 g
Assay Percent Range 97.5% min. (Complexometry)
Linear Formula (CH3CO2)2Cd·2H2O
Merck Index 10,1584
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Formula Weight 266.52
Percent Purity 98%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Cadmium acetate dihydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Very toxic to aquatic life with long lasting effects.
May cause cancer.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a posi

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.