Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium arsenide, 99% (metals basis), Thermo Scientific Chemicals

Catalog No. p-7049786
Click to view available options
Quantity:
10 g

Cadmium arsenide has a electronic properties and is used in 3D crystal structure.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12006-15-4
Molecular Formula As2Cd3
Molecular Weight (g/mol) 487.085
MDL Number MFCD00049723
InChI Key FSIONULHYUVFFA-UHFFFAOYSA-N
Synonym cadmium arsenide
PubChem CID 6391215
SMILES [As]=[Cd].[As]=[Cd].[Cd]

Specifications

Color Gray
Physical Form Lump
Quantity 10 g
Linear Formula Cd3As2
UN Number UN1557
Solubility Information Soluble in HNO3(nitric acid).
Formula Weight 487.04
Percent Purity 99%
Odor Odorless
Density 3.031 g/mL
Assay (metals basis)
Chemical Name or Material Cadmium arsenide
Show More Show Less
Hazard Category H301-H330-H350i
Hazard Statement GHS H Statement
H300-H330-H350
Fatal if swallowed.
Fatal if inhaled.
May cause cancer.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
EINECSNumber 234-484-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.