Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium bromide, 98%, pure, anhydrous

Catalog No. AC453865000
Click to view available options
Quantity:
25 g
100 g
500 g

Catalog No. AC453865000

Supplier: Thermo Scientific Chemicals 453865000

Only null left
Add to Cart

CAS: 7789-42-6 | Br2Cd | 272.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-42-6
Molecular Formula Br2Cd
Molecular Weight (g/mol) 272.22
Synonym cadmium 2+ dibromide, cadmium 2+ ion dibromide, cadmium bromide, anhydrous 25g, cadmium bromide, anhydrous,

Specifications

Boiling Point 963.0°C
Melting Point 566.0°C
Quantity 500 g
Assay Percent Range 97.5% min
Merck Index 15,162
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohols, moderatly soluble in acetone, slightly soluble in ether
Formula Weight 272.22
Percent Purity 98%
Grade Pure
Chemical Name or Material Cadmium bromide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
EINECSNumber 232-165-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.