Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium chloride hemipentahydrate, 99+%, ACROS Organics™

Click to view available options
Quantity:
25g
250g
Packaging:
Glass bottle
7790-78-5
2·5 H2O
228.34
MFCD00799071
7790-78-5
2·5 H2O
228.34
MFCD00799071
Assay Percent Range 79.5 to 81% (CdCl2) (Complexometry)
Linear Formula CdCl2·5/2H2O
Ammonium 50ppm max.
Merck Index 15, 1622
Solubility Information Solubility in water: 1680g/L (20°C). Other solubilities: soluble in acetone, slightly soluble in ethanol and methanol
Formula Weight 228.34
Physical Form Crystalline Powder, Crystals, and/or Chunks
Percent Purity 99+%
Copper (Cu) 5ppm max.
Insoluble in Water 0.005% max.
Insoluble Matter 0.005% max. (in water)
Iron (Fe) 5ppm max.
Lead (Pb) 50ppm max.
Nitrate and Nitrite 0.003% max. (as NO3)
Packaging Glass bottle
Solubility Solubility in water: 1680g/L (20°C). Other solubilities: soluble in acetone, slightly soluble in ethanol and methanol
Sulfate 50ppm max.
Zinc (Zn) 500ppm max.
Color White
Melting Point 34.0°C
Quantity 25g
Chemical Name or Material Cadmium chloride hemipentahydrate, p.a.
Show More Show Less

GHS H Statement
May cause cancer.
May cause genetic defects.
May damage fertility.
May damage the unborn child.
Fatal if inhaled.
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a positi

GHS Signal Word: Danger

RTECSNumber : EV0178000