Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium Chloride, Honeywell™

Catalog Number p-7071863
Click to view available options
Quantity:
25 g
100 g
10108-64-2
CdCl2
183.31
MFCD00010916
YKYOUMDCQGMQQO-UHFFFAOYSA-L
cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
24947
CHEBI:35456
dichlorocadmium
[Cl-].[Cl-].[Cd++]
10108-64-2
CdCl2
183.31
MFCD00010916
YKYOUMDCQGMQQO-UHFFFAOYSA-L
cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
24947
CHEBI:35456
dichlorocadmium
[Cl-].[Cl-].[Cd++]

Puriss. p.a., anhydrous,≥99.0%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY
Physical Form Anhydrous
Quantity 25 g
UN Number 2570
Solubility Information Soluble in water 457g/L at 20°C
Formula Weight 183.32g/mol
Percent Purity ≥99%
Grade puriss. p.a.
Vapor Pressure 10mmHg (656°C)
Density Vapor Density: 6.3 (vs air)
Chemical Name or Material Cadmium Chloride
Hazard Statement P201-P260-P280-P304 + P340 + P310-P308 + P313
Precautionary Statement H301-H330-H340-H350-H360FD-H372-H410
EINECSNumber 233-296-7
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit