Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadmium chloride hydrate, 99.99% (metals basis)
Click to view available options
Quantity:
25 g
100 g
Description
Cadmium chloride hydrate is used in laboratory chemicals and manufacture of substances.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 25 g |
| UN Number | UN2570 |
| Merck Index | 14,1617 |
| Solubility Information | Soluble in water. |
| Formula Weight | 183.32 (Anhydrous) |
| Percent Purity | 99.99% |
| Refractive Index | 1.6513 |
| Sensitivity | Hygroscopic |
| Assay | (metals basis) |
| Show More |
Safety and Handling
| Hazard Category | H301-H330-H340-H350-H360-H372 |
| Hazard Statement | GHS H Statement H300-H330-H340-H350-H360-H372 Fatal if swallowed. Fatal if inhaled. May cause genetic defects. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CADMIUM COMPOUNDS |
| EINECSNumber | 233-296-7 |
| RTECSNumber | EV0175000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction