Learn More
Cadmium chloride, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals
Description
Cadmium chloride is utilized in the preparation of CdTe nanocrystals, highly luminescent. It is also employed in analytical chemistry, photography, dyeing and calico printing, ingredient of electroplating baths, manufacture of special mirrors, the preparation of cadmium standard, analysis of sulfides, and testing pyridine bases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Boiling Point | 960°C |
Melting Point | 568°C |
Physical Form | Beads |
Quantity | 5 g |
UN Number | UN2570 |
Merck Index | 14,1617 |
Solubility Information | Soluble in water and acetone. |
Formula Weight | 183.32 |
Percent Purity | 99.998% |
Odor | Odorless |
Show More |
Safety and Handling
Hazard Category | H301-H330-H335-H340-H350-H360FD-H372 |
Hazard Statement | GHS H Statement H301-H330-H340-H350-H360-H372 Toxic if swallowed. Fatal if inhaled. May cause genetic defects. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. |
Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS |
EINECSNumber | 233-296-7 |
RTECSNumber | EV0175000 |
TSCA | Yes |
storageNote1 |
|
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.