Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium hydroxide, 75%(as Cd), pure, Thermo Scientific™

Catalog No. AC453882500
Click to view available options
Quantity:
250 g
1 kg

Catalog No. AC453882500

Supplier: Thermo Scientific Chemicals 453882500

Chemical Identifiers

CAS 21041-95-2
Molecular Formula CdH2O2
Molecular Weight (g/mol) 146.43
MDL Number MFCD00015995
InChI Key PLLZRTNVEXYBNA-UHFFFAOYSA-L
IUPAC Name cadmium(2+) dihydroxide
SMILES [OH-].[OH-].[Cd++]

Specifications

Melting Point 232.0°C
Quantity 250 g
Assay Percent Range 75%(as Cd)
Merck Index 15,1625
Solubility Information Solubility in water: insoluble. Other solubilities: slightly sol in naoh sol, sol in dil acids, in nh4, or nh4cl sol
Formula Weight 146.41
Percent Purity 75% (as Cd)
Grade Pure
Chemical Name or Material Cadmium hydroxide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if inhaled.
Suspected of causing genetic defects.
May cause cancer.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
Causes damage to organs through prolonged or repeated e
Precautionary Statement GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Use personal protective equipment as required.
EINECSNumber 244-168-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.