Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium hydroxide, Puratronic™, 99.98% (metals basis)

Catalog No. p-7048482
Click to view available options
Quantity:
2 g
10 g
25 g

CAS: 21041-95-2 | CdH2O2 | 146.43 g/mol

It is found in the industrial nickel cadmium storage batteries, in cadmium plating, in the manufacture of cadmium salts, laboratory reagents. It is a useful biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21041-95-2
Molecular Formula CdH2O2
Molecular Weight (g/mol) 146.43
MDL Number MFCD00015995
InChI Key PLLZRTNVEXYBNA-UHFFFAOYSA-L
IUPAC Name cadmium(2+) dihydroxide
SMILES [OH-].[OH-].[Cd++]

Specifications

Melting Point 300°C (decomposition)
Physical Form Powder
Quantity 2 g
UN Number UN2570
Merck Index 14,1620
Solubility Information Soluble in dilute acids. Slightly soluble in water; 0.026g/100 mL.
Formula Weight 146.41
Percent Purity 99.98%
Grade Puratronic™
Sensitivity Air Sensitive
Density 4.79 g/mL at 15°C
Assay (metals basis)
Chemical Name or Material Cadmium hydroxide
Show More Show Less
Hazard Category H330-H341-H350-H361fd-H372
Hazard Statement GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P284-P304+P340-P308+P313-P310-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
EINECSNumber 244-168-5
RTECSNumber EV1260000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.