Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium iodide, 99.5% (metals basis)

Catalog No. p-7049175
Click to view available options
Quantity:
25 g
100 g

CAS: 7790-80-9 | CdI2 | 366.223 g/mol

It is used in lithography, photography, electroplating, as an intermediate and in the manufacturing of phosphors, lubricants, electrode position of Cadmium,

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In photography, process engraving and lithography, analytical chemistry, electrodeposition of Cadmium, manufacturing of phosphors, lubricants
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-80-9
Molecular Formula CdI2
Molecular Weight (g/mol) 366.223
MDL Number MFCD00010920
InChI Key OKIIEJOIXGHUKX-UHFFFAOYSA-L
Synonym cadmium iodide, cadmium diiodide, unii-2f2upu4kcw, cadmium 2+ diiodide, 2f2upu4kcw, acmc-20aljc, cadmium 2+ ion diiodide, cadmium iodide trace metals basis 25g
PubChem CID 24635
IUPAC Name cadmium(2+);diiodide
SMILES [Cd+2].[I-].[I-]

Specifications

Boiling Point 787°C
Melting Point 387°C to 389°C
Color Green-Yellow
Quantity 25 g
UN Number UN2570
Merck Index 14,1621
Solubility Information Soluble in water, alcohol, ether, acetone, ammonia, and acids
Formula Weight 366.21
Percent Purity 99.5%
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Density 5.67 g/mL
Assay (metals basis)
Chemical Name or Material Cadmium iodide
Show More Show Less
Hazard Category H301+H331-H351-H373
Hazard Statement GHS H Statement
H301-H331-H351-H373
Toxic if swallowed.
Toxic if inhaled.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
EINECSNumber 232-223-6
RTECSNumber EV1290000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.