Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cadmium Monoclonal Antibody (3A9)
GREENER_CHOICE

Catalog No. PIMA549724
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549724 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549724 Supplier Invitrogen™ Supplier No. MA549724
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Cadmium is a chemical element with the symbol Cd and atomic number 48. It is a soft, bluish-white metal that is found naturally in the earth's crust. Cadmium is commonly used in batteries, pigments, and coatings, as well as in some industrial processes. However, cadmium is also a toxic metal that can cause harm to human health and the environment. Exposure to high levels of cadmium can lead to lung and prostate cancer, kidney damage, and other health problems.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cadmium
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 3A9
Concentration 10 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias Cd
Host Species Mouse
Immunogen Conjugate of Cd hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.