Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Cadmium Nitrate, Tetrahydrate
SCI_ED

Catalog No. s25214a Shop All Fisherbrand Products
Click to view available options
:
100g

Specifications

Color White
CAS 10022-68-1
Physical Form Solid
Quantity 100 g
Assay Percent Range >98%
Molecular Formula CdH2N2O6
Linear Formula Cd(NO3)2·4H2O
MDL Number MFCD00149626
Synonym cadmium nitrate tetrahydrate, nitric acid, cadmium salt, tetrahydrate, cd no3 2.4h2o, acmc-20aljb, ksc174c1h, cadmium nitrate-water 1/4, cadmiumnitratetetrahydrate, cadmium ii nitrate tetrahydrate, nitric acid, cadmium salt tetrahydrate
InChI Key IZIPRBSCGMISKX-UHFFFAOYSA-N
SMILES [Cd++].O[N+]([O-])=O.O[N+]([O-])=O
IUPAC Name cadmium(2+) bis(nitric acid)
Molecular Weight (g/mol) 238.44
PubChem CID 56924536
ChEBI CHEBI:86156
Formula Weight 308.49
Packaging Solid
Chemical Name or Material Cadmium Nitrate, Tetrahydrate
Show More Show Less
ShelfLife 5 years
DOTInformation DOT Class 5.1, II, : Oxidizer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.