Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium selenide, 99.995% (metals basis)

Catalog No. AA3632306
Click to view available options
Quantity:
5 g
25 g
100 g

Catalog No. AA3632306

Supplier: Thermo Scientific Chemicals 03632306

Only null left
Add to Cart

Vacuum deposition | CAS: 1306-24-7 | CdSe | 191.39 g/mol

It ia a photoconductive and semiconductor material used in a cell where a fast response time and high sensitivity to longer wavelengths of light is required; also used as a red pigment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1306-24-7
Molecular Formula CdSe
Molecular Weight (g/mol) 191.39
MDL Number MFCD00010917
InChI Key AQCDIIAORKRFCD-UHFFFAOYSA-N
Synonym cadmium selenide, cadmium ii selenide, cadmium 2+ selenide, cadmium selenide nanotubes, cadmium selenide trace metals basis 1g, cadmium selenide candot quantum dot core , 50umol/l in hexane, lumidot tm cdse, 480, core-type quantum dots, 5 mg/ml in toluene, lumidot tm cdse, 520, core-type quantum dots, 5 mg/ml in toluene
PubChem CID 14784
ChEBI CHEBI:50834
IUPAC Name selanylidenecadmium
SMILES [Se]=[Cd]

Specifications

Melting Point >1350°C
Color Black to Brown-Red
Physical Form Sintered Lump
Quantity 5 g
UN Number UN3077
Merck Index 14,1626
Solubility Information Insoluble in water but soluble in acids.
Formula Weight 191.36
Percent Purity 99.995%
Odor Odorless
Density 5.81 g/mL
Assay (metals basis)
Chemical Name or Material Cadmium selenide
Show More Show Less
Hazard Category H301+H331-H350-H373
Hazard Statement GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 215-148-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.