Learn More
Cadmium selenide, 99.999% (metals basis)
Description
Cadmium selenide is applied in opto-electronic devices, laser diodes, biomedical imaging, nanosensing, high-efficiency solar cells and thin-film transistors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | >1350°C |
| Color | Black to Brown-Red |
| Physical Form | Pieces |
| Quantity | 50 g |
| UN Number | UN3077 |
| Merck Index | 14,1626 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 191.36 |
| Percent Purity | 99.999% |
| Odor | Odorless |
| Show More |
Safety and Handling
| Hazard Category | H301+H331-H350-H373 |
| Hazard Statement | GHS H Statement H301-H331-H373 Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c |
| DOTInformation | Hazard Class: 6.1; Packaging Group: III |
| EINECSNumber | 215-148-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.