Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium Sulfate 8/3-Hydrate, puriss. p.a. ACS Reagent, ≥99.0%, Solstice

Catalog No. 60045183 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-183 100 g Poly Bottle
60-045-187 25 g Poly Bottle
60-045-188 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-045-183 Supplier Solstice Supplier No. 20920100G

Puriss. p.a., ACS Reagent,≥99.0% (calc. based on CdSO4 · 8/3H2O, KT)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-84-3
MDL Number MFCD02253000

Specifications

Melting Point 41.5°C
Color Colorless
Physical Form Solid
Quantity 100 g
Linear Formula CdSO4 · 8/3H2O
UN Number UN2570
Formula Weight 256.57g/mol
Percent Purity ≥99.0% (calc. based on CdSO4.83H2O, KT)
Grade ACS/puriss. p.a.
Packaging Poly Bottle
Density 3.09g/cu.cm
Chemical Name or Material Cadmium Sulfate 8/3-Hydrate
Show More Show Less
Hazard Statement P201-P260-P301 + P310 + P330-P304 + P340 + P310-P308 + P313-P403 + P233
Precautionary Statement H301-H330-H340-H350-H360FD-H372-H410
DOTInformation class 6.1 - PG 3 - EHS - Cadmium compounds, HI: all (not BR)
EINECSNumber 233-331-6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.