Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium sulfate, anhydrous, ACS, 99+%

Catalog No. AA1186122
Click to view available options
Quantity:
25 g
100 g

CAS: 10124-36-4 | CdO4S | 208.47 g/mol

Cadmium sulfate is used in pigments and electroplating. It acts as an accelerator in cement formation. It is also employed as lubricants. Further, it is useful for the electroplating of cadmium in electronic circuits. It serves as a precursor to prepare cadmium-based pigment like cadmium sulfide. In the Western standard cell, it is used for electrolyte.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10124-36-4
Molecular Formula CdO4S
Molecular Weight (g/mol) 208.47
MDL Number MFCD00010923
InChI Key QCUOBSQYDGUHHT-UHFFFAOYSA-L
Synonym cadmium sulfate, cadmium sulphate, cadmium monosulfate, sulfuric acid, cadmium salt 1:1, cadmium sulfuricum, cadmium sulfate 1:1, cadmium sulfate, tetrahydrate, cadmium 2+ sulfate, caswell no. 136c, unii-947unf3z6o
PubChem CID 24962
ChEBI CHEBI:50292
IUPAC Name cadmium(2+);sulfate
SMILES [O-]S(=O)(=O)[O-].[Cd+2]

Specifications

Melting Point 1000°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range >99%
Solubility Information Soluble in water. Slightly soluble in methanol and ethyl acetate. Insoluble in ethanol.
Formula Weight 208.46
Grade ACS
Sensitivity Hygroscopic
Appearance White powder
Density 4.691 g/cm3
Chemical Name or Material Cadmium sulfate, For analysis ACS
Show More Show Less
Hazard Category H301-H330-H340-H350-H360FD-H372
Precautionary Statement P201-P202-P260-P264b-P270-P271-P280-P301+P310-P304+P340-P308+P313-P310-P330-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 233-331-6
RTECSNumber EV2700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.