Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium sulfide, 99.999% (metals basis)

Catalog No. p-7050449
Click to view available options
Quantity:
50 g
250 g

CAS: 1306-23-6 | CdS | 144.47 g/mol

The main application of cadmium sulfide is as a pigment. Cadmium sulfide is also used in the production of solar cells. Used as light dependent resistors/photo resistors for light sensors. Used in MOCVD, sputtering and electrochemical deposition. In screen printing, it is used as a slurry containing dispersed cadmium sulfide. It is also used in dandruff shampoos.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1306-23-6
Molecular Formula CdS
Molecular Weight (g/mol) 144.47
MDL Number MFCD00010922
InChI Key FRLJSGOEGLARCA-UHFFFAOYSA-N
Synonym cadmium sulfide, cadmium sulphide, capsebon, orange cadmium, cadmopur yellow, jaune brilliant, aurora yellow, cadmium orange, cadmium yellow, ferro yellow
PubChem CID 14783
ChEBI CHEBI:50833
IUPAC Name cadmium(2+) sulfanediide
SMILES [S--].[Cd++]

Specifications

Melting Point 980°C (sublimation)
Color Orange
Physical Form Powder/Granules
Quantity 50 g
UN Number UN3077
Merck Index 14,1628
Solubility Information Soluble in acid,very slightly soluble in ammonium hydroxide.
Formula Weight 144.46
Percent Purity 99.999%
Odor Odorless
Density 4.82 g/mL
Assay (metals basis)
Chemical Name or Material Cadmium sulfide
Show More Show Less
Hazard Category H302-H341-H350-H361fd-H372
Hazard Statement GHS H Statement
H350-H372-H341-H361-H302
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Precautionary Statement P201-P202-P260-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 215-147-8
RTECSNumber EV3150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.