Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium telluride, Black polycrystalline lump, 99.999% (Metals basis), Thermo Scientific Chemicals

Catalog No. p-4824649
Click to view available options
Quantity:
25 g
100 g

Cadmium telluride finds applications as the semiconductor material in photovoltaics and infrared optical window. It is used in the production of thin film solar cells. It is doped with chlorine and used as a radiation detector for x-rays, alfa and beta particles. .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1306-25-8
Molecular Formula CdTe
Molecular Weight (g/mol) 240.01
MDL Number MFCD00015998
InChI Key RPPBZEBXAAZZJH-UHFFFAOYSA-N
Synonym cadmium telluride, irtran 6, cadmium monotelluride, unii-stg188wo13, cadmiumtellurid, irtran-6, cadmium telluride metals basis, cadmium telluride trace metals basis
PubChem CID 91501
IUPAC Name tellanylidenecadmium
SMILES [Cd]=[Te]

Specifications

Melting Point 1041°C
Color Black
Physical Form Polycrystalline Lump
Quantity 25 g
Assay Percent Range 99.999% (Metals basis)
Solubility Information Insoluble in water.
Formula Weight 240
Odor Odorless
Density 6.2
Chemical Name or Material Cadmium telluride
Hazard Category H332-H350-H372
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P304+P340-P308+P313-P501c
DOTInformation DOT Class: 9, Packaging Group: III
EINECSNumber 215-149-9
RTECSNumber EV3330000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.