Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAF-1 p150 Monoclonal Antibody (1), Active Motif™
SDP

Mouse Monoclonal Antibody

$135.30 - $405.90

Specifications

Antigen CAF-1 p150
Clone 1
Concentration 1 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3028
SDP
Encompass
View Documents
Active Motif
39998
10 μg
Each for $135.30
Only null left
Add to Cart
 
50-199-3027
SDP
Encompass
View Documents
Active Motif
39997
100 μg
Each for $405.90
Only null left
Add to Cart
 
Description

Description

Chromatin assembly factor I (CAF1) is a nuclear complex consisting of p50, p60 (CHAF1B), and p150 (CHAF1A) subunits that assembles histone octamers onto replicating DNA in vitro.
Specifications

Specifications

CAF-1 p150
1 mg/mL
Monoclonal
Liquid
RUO
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
AL023013; AL024058; Cac1p; CAF; CAF1; CAF-1; CAF-1 subunit A; CAF1B; CAF1P150; CAF-I 150 kDa subunit; CAF-I p150; CAF-Ip150; Caip150; CHAF1A; Chromatin assembly factor 1 subunit A; chromatin assembly factor 1, subunit A (p150); chromatin assembly factor I (150 kDa); Chromatin assembly factor I p150 subunit; CTB-50L17.7; hp150; MGC71229; P150
CHAF1A
IgG1
Protein G
Antibody
1
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
Mouse
Human
Q13111
10036
The CAF-1 p150 antobody was raised against full-length CAF-1 purified from human 293 cells.
Primary
-20°C, Avoid Freeze/Thaw Cycles
CHAF1A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.