Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAF1 p150 Polyclonal Antibody
Supplier: Thermo Scientific PA5116964
Description

Specifications
CAF1 p150 | |
Polyclonal | |
Unconjugated | |
CHAF1A | |
AL023013; AL024058; Cac1p; CAF; CAF1; CAF-1; CAF-1 subunit A; CAF1B; CAF1P150; CAF-I 150 kDa subunit; CAF-I p150; CAF-Ip150; Caip150; CHAF1A; Chromatin assembly factor 1 subunit A; chromatin assembly factor 1, subunit A (p150); chromatin assembly factor I (150 kDa); chromatin assembly factor I p150 subunit; CTB-50L17.7; hp150; MGC71229; P150 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10036 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.98 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q13111 | |
CHAF1A | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human p150 CAF1 (NP_0054742). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction