Learn More
Invitrogen™ CAF1 p150 Recombinant Rabbit Monoclonal Antibody (JG38-98)

Description
Positive Control: K562, A431, 293T, LOVO, human tonsil tissue, human colon tissue, Daudi.
Specifications
Specifications
| Antigen | CAF1 p150 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | JG38-98 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | CHAF1A |
| Gene Accession No. | Q13111 |
| Gene Alias | AL023013; AL024058; Cac1p; CAF; CAF1; CAF-1; CAF-1 subunit A; CAF1B; CAF1P150; CAF-I 150 kDa subunit; CAF-I p150; CAF-Ip150; Caip150; CHAF1A; Chromatin assembly factor 1 subunit A; chromatin assembly factor 1, subunit A (p150); chromatin assembly factor I (150 kDa); chromatin assembly factor I p150 subunit; CTB-50L17.7; hp150; MGC71229; P150 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.