Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAF1 p60 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575333
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575333 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575333 Supplier Invitrogen™ Supplier No. PA575333
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Chromatin assembly factor 1 (CAF-1) is a member of the histone chaperone family. It is required for inheritance of epigenetically determined chromosomal states, and in vitro, it promotes the assembly of chromatin during DNA replication and DNA repair. CAF-1 is a complex of three subunits; p150, p60, and p48. This complex is localized to the nucleus of human cells. Both the composition and the biochemical activity of CAF-1 are conserved among human, yeast, and Drosophila.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAF1 p60
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Chaf1b
Gene Accession No. Q13112, Q9D0N7
Gene Alias 2600017H24Rik; C76145; CAF1; CAF-1; CAF1 p60; CAF-1 subunit B; CAF1A; CAF1P60; CAF-I 60 kDa subunit; CAF-I p60; CAF-Ip60; CHAF1B; Chromatin assembly factor 1 subunit B; chromatin assembly factor 1, subunit B; chromatin assembly factor 1, subunit B (p60); chromatin assembly factor I p60 subunit; fe36g06; gef; human chromatin assembly factor-I p60 subunit; LOW QUALITY PROTEIN: chromatin assembly factor 1 subunit B; M-phase phosphoprotein 7; MPHOSPH7; MPP7; p60 CAF1; p60-CAF1; wu:fd07c09; wu:fe36g06; wu:fv38g09; zgc:56096
Gene Symbols Chaf1b
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 70-120 of Human CAF1 p60.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110749, 288242, 8208
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.