Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAGE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550924
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550924 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550924 Supplier Invitrogen™ Supplier No. PA550924
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CAGE1 Polyclonal Antibody detects endogenous levels of CAGE1 protein.

CAGE1 (Cancer Antigen 1) is a Protein Coding gene. Diseases associated with CAGE1 include Fat Necrosis Of Breast and Amelogenesis Imperfecta, Type If. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAGE1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CAGE1
Gene Accession No. Q8TC20
Gene Alias 4933427I01Rik; bA69L16.7; CAGE1; CAGE-1; cancer antigen 1; cancer/testis antigen 3; Cancer/testis antigen 3 homolog; cancer/testis antigen 95; cancer/testis antigen gene 1; cancer-associated gene 1 protein; Cancer-associated gene 1 protein homolog; CT3; CT3 homolog; CT95; Ctag3
Gene Symbols CAGE1
Host Species Rabbit
Immunogen Synthesized peptide derived from the C-terminal region of human CAGE-1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285782
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.