Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAGE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106626
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106626 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106626 Supplier Invitrogen™ Supplier No. PA5106626
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CAGE1.

CAGE1 (Cancer Antigen 1) is a Protein Coding gene. Diseases associated with CAGE1 include Fat Necrosis Of Breast and Amelogenesis Imperfecta, Type If. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAGE1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CAGE1
Gene Accession No. Q5IR70, Q66HB6, Q8TC20
Gene Alias 4933427I01Rik; bA69L16.7; CAGE1; CAGE-1; cancer antigen 1; cancer/testis antigen 3; Cancer/testis antigen 3 homolog; cancer/testis antigen 95; cancer/testis antigen gene 1; cancer-associated gene 1 protein; Cancer-associated gene 1 protein homolog; CT3; CT3 homolog; CT95; Ctag3
Gene Symbols CAGE1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CAGE1(Accession Q8TC20), corresponding to amino acid residues A720-K770.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285782, 306872, 71213
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.