Learn More
MilliporeSigma™ Supelco™ Cages for Fused Silica Columns
Reduce the possibility of damaging the protective coating on columns; design makes it easy to hang columns in most gas chromatography ovens
$119.00
Specifications
For Use With (Application) | Hanging Columns in Gas Chromatography Ovens |
---|---|
Quantity | 3/Pk. |
Type | Fused Silica Column Cages |
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
11-100-4750
|
MilliporeSigma™ Supelco™
23742 |
0.25 mm I.D. Columns |
Each of 1 for $119.00
|
|
|||||
Description
- The ends of flexible fused-silica capillary tubing are intrinsically straight and therefore the column must be coiled and properly confined on a circular frame
- This allows for the capillary column to then be safely mounted in the column oven of the gas chromatograph
- A unique design with minimal metal-to-column contact reduces the possibility of damaging the protective coating on columns
- Simplifies the process of hanging columns in most GC ovens
Specifications
Hanging Columns in Gas Chromatography Ovens | |
3/Pk. | |
Fused Silica Column Cages |