Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CaI-PLA2 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS6709R

Description
The protein encoded by this gene is an A2 phospholipase, a class of enzyme that catalyzes the release of fatty acids from phospholipids. The encoded protein may play a role in phospholipid remodelling, arachidonic acid release, leukotriene and prostaglandin synthesis, fas-mediated apoptosis, and transmembrane ion flux in glucose-stimulated B-cells. Several transcript variants encoding multiple isoforms have been described, but the full-length nature of only two of them have been determined to date.Specifications
CaI-PLA2 | |
Polyclonal | |
Unconjugated | |
PLA2G6 | |
85 kDa calcium-independent phospholipase A2; 85/88 kDa calcium-independent phospholipase A2; BB112799; CaI-PLA2; Group VI phospholipase A2; GVI; GVI PLA2; INAD1; Intracellular membrane-associated calcium-independent phospholipase A2 beta; iPLA(2)beta; iPLA2; iPLA2beta; iPLA2-beta; IPLA2-VIA; NBIA2; NBIA2A; NBIA2B; neurodegeneration with brain iron accumulation 2; PARK14; Patatin-like phospholipase domain-containing protein 9; phospholipase A2 group VI; phospholipase A2, group VI; phospholipase A2, group VI (cytosolic, calcium-independent); PLA2; Pla2g6; PLPLA9; Pnpla9 | |
Rabbit | |
Protein A | |
RUO | |
8398 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
O60733 | |
PLA2G6 | |
KLH conjugated synthetic peptide derived from human CaI-PLA2/PLA2G6. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction