Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Streck Cal-Chex™ Calibrator

Click to view available options
Quantity:
1 x 3 mL
3 x 3 mL
Description
- Assayed on Beckman Coulter, Horiba Medical, Siemens and Mindray instruments.
- Plastic vials with pierceable caps for analyzer autosampling.
- 5-day open-vial stability, 45-day closed-vial stability.
- Established assay ranges provide less room for error.
- Helps clinical laboratories meet quality requirements.
Order Info
3.0mL vial has pierceable cap
Specifications
Specifications
Content And Storage | 2°C to 10°C |
Type | Cal-Chex Hematology Calibrators |
Intended Use | Whole Blood calibration |
Quantity | 3 x 3 mL |
Stability | 5 Days Open, 45 Days Closed Vial |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction