Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CALB2 Mouse anti-Human, Clone:5E9B6, Abnova™

Catalog No. 89333157 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-157 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-157 Supplier Abnova Corporation Supplier No. MAB16707
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human CALB2.

This gene encodes an intracellular calcium-binding protein belonging to the troponin C superfamily. Members of this protein family have six EF-hand domains which bind calcium. Three alternatively spliced transcript variants that encode different proteins have been described. [provided by RefSeq]

Specifications

Antigen CALB2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5E9B6
Conjugate Unconjugated
Description calbindin 2
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry
Immunocytochemistry
Flow Cytometry
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene CALB2
Gene Alias CAL2
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 1-271 of human CALB2 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 794
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.