Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calbindin/Calretinin Recombinant Rabbit Monoclonal Antibody (HL1787)
GREENER_CHOICE

Catalog No. FEMA550334
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550334 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550334 Supplier Invitrogen™ Supplier No. MA550334
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Calbindin/Calretinin Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (F), IHC (P)

Calbindin/Calretinin is a gene target that codes for two calcium-binding proteins that are expressed in different regions of the nervous system. Calbindin is mainly found in the cerebellum, while calretinin is expressed in the hippocampus, cortex, and other brain regions. These proteins play an essential role in calcium signaling, which is crucial for neuronal function and synaptic plasticity. They are also involved in other cellular processes, such as cell survival, differentiation, and gene expression. Dysregulation of calbindin/calretinin expression has been linked to various neurological disorders, including epilepsy, Alzheimer's disease, and Parkinson's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calbindin/Calretinin
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1787
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CALB2
Gene Accession No. P05937, P07171, P12658, P22676, P47728, Q08331
Gene Alias CALB1; CALB2
Gene Symbols CALB1, CALB2
Host Species Rabbit
Immunogen Recombinant fragment of human Calbindin.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117059, 12307, 12308, 793, 794, 83839
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.