Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calbindin D-28K Antibody (CALB1/2364), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308309B
Description
Calbindin D-28K Monoclonal antibody specifically detects Calbindin D-28K in Human samples. It is validated for ELISA, Protein ArraySpecifications
Calbindin D-28K | |
Monoclonal | |
Biotin | |
CAB27, CALB, calbindin, calbindin 1, (28kD), calbindin 1, 28kDa, Calbindin D28, D-28K, RTVL-H protein, Vitamin D-dependent calcium-binding protein, avian-type | |
Recombinant fragment (around aa 7-96) of human Calbindin D-28K protein (exact sequence is proprietary) (Uniprot: P05937) | |
100 μg | |
Neuronal Cell Markers, Neuroscience, Neurotransmission, Stem Cell Markers | |
793 | |
Store at 4°C in the dark. | |
IgG1 κ |
ELISA, Protein Array | |
CALB1/2364 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction