Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calbindin D-28K Rabbit anti-Mouse, Human, Rat, Polyclonal, MilliporeSigma™

Catalog No. ABN219225UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN2192-25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN2192-25UL Supplier MilliporeSigma Supplier No. ABN219225UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Calbindin D-28K Antibody is a highly specific rabbit polyclonal antibody that targets Calbindin D and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calbindin D-28K
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Calb1
Host Species Rabbit
Immunogen GST/His-tagged recombinant fragment corresponding to 107 amino acids from the C-terminal end of murine Calbindin D-28K.
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NM_004929.2
Test Specificity This rabbit polyclonal antibody detects Calbindin D-28K in Human, Murine, and Rat cells. It targets an epitope within 107 amino acids from the C-terminal region.
Target Species Mouse, Human, Rat
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.