Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calbindin D28K Monoclonal Antibody (4H7)

Catalog No. PIMA547339
Change view
Click to view available options
Quantity:
100 μL
500 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547339 500 μL
PIMA547438 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA547339 Supplier Invitrogen™ Supplier No. MA547339
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calbindin D28K Monoclonal Antibody for Western Blot, ICC/IF, IHC (PFA)

Calbindin-D-28K (also termed vitamin D-dependent calcium-binding protein, or cholecalcin), is a highly conserved 28kDa calcium binding protein, with broad tissue distribution. It belongs, together with calmodulin, S-100, parvalbumin, troponin C and other proteins, to a family of low molecular weight calcium-binding proteins (CaBPs). These CaBPs have homologous primary structures, which contain conserved polypeptide folds of the EF-hand type for Ca2+ binding. Calbindin-D-28K is found predominantly in subpopulations of central and peripheral nervous system neurons, and in certain epithelial nervous system neurons, and in certain epithelial cells involved in Ca2+ transport such as distal tubular cells and cortical collecting tubules of the kidney, and in enteric neuroendocrine cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calbindin D28K
Applications Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4H7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 5mM sodium azide
Gene CALB1
Gene Accession No. P04467, P05937, P07171, P12658
Gene Alias Brain-2; CAB27; CaBP28K; Calb; CALB1; Calb-1; Calbindin; calbindin 1; calbindin 1, (28kD); calbindin 1, 28 kD; calbindin 1, 28kDa; calbindin D28; calbindin-28K; calbindin-D (28k); CalbindinD28K; CB; cerebellar Ca-binding protein spot 35 protein; cerebellar Ca-binding protein, spot 35 protein; D-28K; PCD-29; RTVL-H protein; spot 35 protein; Vitamin D-dependent calcium-binding protein, avian-type
Gene Symbols CALB1
Host Species Mouse
Immunogen Full-length recombinant human calbindin protein expressed in and purified from E. coli.
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100153645, 12307, 539930, 793, 83839
Target Species Bovine, Human, Mouse, Pig, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.